Assay Details
| Gene Symbol: SLC25A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284446.1 | NP_001271375.1 | 8 - 9 | 892 | 127 | |
| XM_005641802.2 | XP_005641859.1 | 9 - 10 | 1028 | 127 | ||
| XM_005641803.3 | XP_005641860.1 | 9 - 10 | 1066 | 127 | ||
| XM_014111352.2 | XP_013966827.1 | 9 - 10 | 1157 | 127 | ||
| XM_847404.5 | XP_852497.1 | 9 - 10 | 1166 | 127 | ||
| GenBank mRNA | AB612119.1 | - | 8 - 9 | 892 | 127 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC25A14
Genomic Map