Assay Details
| Gene Symbol: PAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637622.3 | XP_005637679.1 | 4 - 5 | 487 | 147 | |
| XM_005637623.3 | XP_005637680.1 | 4 - 5 | 484 | 147 | ||
| XM_014109102.2 | XP_013964577.1 | 3 - 4 | 3780 | 147 | ||
| XM_014109103.2 | XP_013964578.1 | 3 - 4 | 3773 | 147 | ||
| XM_014109104.2 | XP_013964579.1 | 4 - 5 | 487 | 147 | ||
| XM_014109105.2 | XP_013964580.1 | 4 - 5 | 484 | 147 | ||
More Information
Set Membership: |
|

PAX2
Genomic Map