Assay Details
| Gene Symbol: LOC610380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624197.3 | XP_005624254.1 | 724 | 65 | ||
| XM_014116365.2 | XP_013971840.1 | 938 | 65 | |||
| XM_847872.5 | XP_852965.1 | 941 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC610380
Genomic Map