Assay Details
| Gene Symbol: NAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629165.1 | XP_005629222.1 | 6 - 7 | 470 | 104 | |
| XM_022428108.1 | XP_022283816.1 | 6 - 7 | 576 | 104 | ||
| XM_022428109.1 | XP_022283817.1 | 6 - 7 | 576 | 104 | ||
| XM_022428110.1 | XP_022283818.1 | 6 - 7 | 576 | 104 | ||
| XM_022428111.1 | XP_022283819.1 | 7 - 8 | 601 | 104 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NAP1L1
Genomic Map