Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113765.2 | XP_013969240.1 | 3 - 4 | 368 | 144 | |
| XM_848083.5 | XP_853176.2 | 4 - 5 | 529 | 144 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TTC4
Genomic Map