Assay Details
| Gene Symbol: B4GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421507.1 | XP_022277215.1 | 6 - 7 | 803 | 125 | |
| XM_547621.6 | XP_547621.2 | 5 - 6 | 721 | 125 | ||
More Information
Set Membership: |
|

B4GALT6
Genomic Map