Assay Details
| Gene Symbol: LOC490812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422274.1 | XP_022277982.1 | 11 - 12 | 3319 | 146 | |
| XM_022422275.1 | XP_022277983.1 | 10 - 11 | 3307 | 146 | ||
| XM_022422276.1 | XP_022277984.1 | 10 - 11 | 3294 | 146 | ||
More Information
Set Membership: |
|

LOC490812
Genomic Map