Assay Details
| Gene Symbol: LOC490812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422274.1 | XP_022277982.1 | 15 - 16 | 4042 | 156 | |
| XM_022422275.1 | XP_022277983.1 | 14 - 15 | 4030 | 156 | ||
| XM_022422276.1 | XP_022277984.1 | 14 - 15 | 4017 | 156 | ||
| XM_022422277.1 | XP_022277985.1 | 14 - 15 | 4015 | 156 | ||
More Information
Set Membership: |
|

LOC490812
Genomic Map