Assay Details
| Gene Symbol: EI24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418130.1 | XP_022273838.1 | 1 | 925 | 105 | |
| XM_546419.5 | XP_546419.1 | 1 | 938 | 105 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EI24
Genomic Map