Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619912.3 | XP_005619969.1 | 6 - 7 | 651 | 168 | |
| XM_005619913.2 | XP_005619970.1 | 6 - 7 | 906 | 168 | ||
| XM_022418448.1 | XP_022274156.1 | 6 - 7 | 765 | 168 | ||
| XM_022418449.1 | XP_022274157.1 | 6 - 7 | 1021 | 168 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPAG7
Genomic Map