Assay Details
| Gene Symbol: HNRNPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637504.2 | XP_005637561.1 | 4 | 545 | 65 | |
| XM_005637505.2 | XP_005637562.1 | 3 | 480 | 65 | ||
| XM_022411808.1 | XP_022267516.1 | 5 | 683 | 65 | ||
| XM_843604.5 | XP_848697.1 | 3 | 532 | 65 | ||
| XM_851801.5 | XP_856894.1 | 4 | 655 | 65 | ||
| XM_851880.5 | XP_856973.1 | 2 | 438 | 65 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HNRNPF
Genomic Map