Assay Details
| Gene Symbol: FXYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195142.1 | NP_001182071.1 | 5 - 6 | 326 | 114 | |
| NM_001195143.1 | NP_001182072.1 | 5 - 6 | 326 | 114 | ||
| XM_005615754.3 | XP_005615811.2 | 5 - 6 | 379 | 114 | ||
| XM_005615755.3 | XP_005615812.2 | 5 - 6 | 435 | 189 | ||
| XM_014114177.2 | XP_013969652.2 | 5 - 6 | 620 | 189 | ||
| XM_014114183.2 | XP_013969658.1 | 5 - 6 | 530 | 189 | ||
| XM_014114188.2 | XP_013969663.1 | 5 - 6 | 481 | 189 | ||
| GenBank mRNA | M63934.1 | - | 5 - 6 | 303 | 114 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FXYD1
Genomic Map