Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111093.2 | XP_013966568.1 | 8 - 9 | 1345 | 150 | |
| XM_536047.5 | XP_536047.2 | 8 - 9 | 1251 | 150 | ||
| GenBank mRNA | DQ403086.1 | - | 6 - 7 | 891 | 150 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IDH1
Genomic Map