Assay Details
| Gene Symbol: IRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639784.4 | XP_003639832.2 | 12 - 13 | 1960 | 116 | |
| XM_005633670.2 | XP_005633727.1 | 8 - 9 | 1185 | 116 | ||
| XM_014122566.2 | XP_013978041.2 | 12 - 13 | 1958 | 116 | ||
| XM_022407838.1 | XP_022263546.1 | 12 - 13 | 1613 | 116 | ||
| XM_022407839.1 | XP_022263547.1 | 12 - 13 | 1963 | 116 | ||
More Information
Set Membership: |
|

IRAG1
Genomic Map