Assay Details
| Gene Symbol: ARID5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630383.3 | XP_005630440.1 | 4 - 5 | 334 | 119 | |
| XM_022404674.1 | XP_022260382.1 | 5 - 6 | 1744 | 119 | ||
| XM_022404675.1 | XP_022260383.1 | 6 - 7 | 944 | 119 | ||
| XM_022404676.1 | XP_022260384.1 | 3 - 4 | 1552 | 119 | ||
| XM_022404677.1 | XP_022260385.1 | 3 - 4 | 310 | 119 | ||
| XM_022404678.1 | XP_022260386.1 | 5 - 6 | 713 | 119 | ||
| XM_540165.6 | XP_540165.2 | 5 - 6 | 502 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ARID5A
Genomic Map