Assay Details
| Gene Symbol: HIVEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417638.1 | XP_022273346.1 | 7201 | 147 | ||
| XM_845203.5 | XP_850296.3 | 7418 | 147 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HIVEP2
Genomic Map