Assay Details
| Gene Symbol: METAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629238.3 | XP_005629295.1 | 8 - 9 | 1042 | 179 | |
| XM_005629239.3 | XP_005629296.1 | 7 - 8 | 938 | 179 | ||
| XM_022403342.1 | XP_022259050.1 | 8 - 9 | 937 | 179 | ||
| XM_861061.4 | XP_866154.2 | 8 - 9 | 965 | 179 | ||
More Information
Set Membership: |
|

METAP2
Genomic Map