Assay Details
| Gene Symbol: DUSP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624787.3 | XP_005624844.1 | 3 | 1194 | 102 | |
| XM_022423280.1 | XP_022278988.1 | 4 | 1326 | 102 | ||
| XM_022423281.1 | XP_022278989.1 | 4 | 1309 | 102 | ||
| XM_022423282.1 | XP_022278990.1 | 3 | 734 | 102 | ||
| XM_548251.6 | XP_548251.1 | 3 | 1214 | 102 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DUSP14
Genomic Map