Assay Details
| Gene Symbol: PITPNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422852.1 | XP_022278560.1 | 975 | 71 | ||
| XM_022422853.1 | XP_022278561.1 | 774 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PITPNC1| Gene Symbol: PITPNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422852.1 | XP_022278560.1 | 975 | 71 | ||
| XM_022422853.1 | XP_022278561.1 | 774 | 71 | |||
Set Membership: |
|