Assay Details
| Gene Symbol: GREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638201.3 | XP_005638258.1 | 2 | 439 | 105 | |
| XM_022412778.1 | XP_022268486.1 | 3 | 469 | 105 | ||
More Information
Set Membership: |
|

GREM1
Genomic Map