Assay Details
| Gene Symbol: PBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415304.1 | XP_022271012.1 | 6 - 7 | 1158 | 80 | |
| XM_022415305.1 | XP_022271013.1 | 5 - 6 | 1376 | 80 | ||
| XM_022415306.1 | XP_022271014.1 | 4 - 5 | 669 | 80 | ||
| XM_022415307.1 | XP_022271015.1 | 4 - 5 | 612 | 80 | ||
| XM_022415308.1 | XP_022271016.1 | 5 - 6 | 1375 | 80 | ||
| XM_022415309.1 | XP_022271017.1 | 3 - 4 | 597 | 80 | ||
| XM_854637.5 | XP_859730.2 | 5 - 6 | 1375 | 80 | ||
More Information
Set Membership: |
|

PBX1
Genomic Map