Assay Details
| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120582.2 | XP_013976057.1 | 3 - 4 | 545 | 147 | |
| XM_022404929.1 | XP_022260637.1 | 2 - 3 | 236 | 147 | ||
| XM_533031.6 | XP_533031.2 | 2 - 3 | 476 | 147 | ||
More Information
Set Membership: |
|

PDZK1
Genomic Map