Assay Details
| Gene Symbol: PIGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622518.3 | XP_005622575.1 | 2 | 443 | 121 | |
| XM_014115183.2 | XP_013970658.1 | 2 | 705 | 121 | ||
| XM_022421057.1 | XP_022276765.1 | 2 | 496 | 121 | ||
| XM_537194.7 | XP_537194.1 | 2 | 488 | 121 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PIGC
Genomic Map