Assay Details
| Gene Symbol: PPP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419889.1 | XP_022275597.1 | 6 - 7 | 606 | 151 | |
| XM_547067.6 | XP_547067.1 | 6 - 7 | 664 | 151 | ||
More Information
Set Membership: |
|

PPP4C
Genomic Map