Assay Details
| Gene Symbol: PTHLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003303.1 | NP_001003303.1 | 3 | 430 | 141 | |
| XM_022410845.1 | XP_022266553.1 | 3 | 1003 | 141 | ||
| XM_022410846.1 | XP_022266554.1 | 4 | 888 | 141 | ||
| XM_022410847.1 | XP_022266555.1 | 3 | 837 | 141 | ||
| XM_022410848.1 | XP_022266556.1 | 4 | 722 | 141 | ||
| XM_022410849.1 | XP_022266557.1 | 3 | 621 | 141 | ||
| GenBank mRNA | U15593.1 | - | 3 | 430 | 141 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTHLH
Genomic Map