Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629576.2 | XP_005629633.1 | 1 | 405 | 198 | |
| XM_532732.5 | XP_532732.2 | 1 | 445 | 198 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTN
Genomic Map