Assay Details
| Gene Symbol: REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626129.3 | XP_005626186.1 | 3 | 496 | 171 | |
| XM_005626130.3 | XP_005626187.1 | 3 | 496 | 171 | ||
| XM_005626131.3 | XP_005626188.1 | 3 | 400 | 171 | ||
| XM_005626132.2 | XP_005626189.1 | 3 | 367 | 171 | ||
More Information
Set Membership: |
|

REL
Genomic Map