Assay Details
| Gene Symbol: ST3GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627975.3 | XP_005628032.1 | 5 - 6 | 1271 | 161 | |
| XM_014118688.2 | XP_013974163.1 | 6 - 7 | 1952 | 161 | ||
| XM_022426722.1 | XP_022282430.1 | 5 - 6 | 1853 | 161 | ||
| XM_022426723.1 | XP_022282431.1 | 6 - 7 | 2005 | 161 | ||
| XM_022426724.1 | XP_022282432.1 | 6 - 7 | 2001 | 161 | ||
| XM_022426725.1 | XP_022282433.1 | 6 - 7 | 1940 | 161 | ||
| XM_022426726.1 | XP_022282434.1 | 4 - 5 | 1798 | 161 | ||
| XM_539171.4 | XP_539171.2 | 3 - 4 | 1270 | 161 | ||
More Information
Set Membership: |
|

ST3GAL1
Genomic Map