Assay Details
| Gene Symbol: SLC5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007142.2 | NP_001007143.2 | 910 | 140 | ||
| XM_005621226.3 | XP_005621283.1 | 1068 | 140 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC5A2
Genomic Map