Assay Details
| Gene Symbol: SRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628477.3 | XP_005628534.1 | 2 - 3 | 398 | 177 | |
| XM_532452.5 | XP_532452.1 | 2 - 3 | 212 | 177 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRI
Genomic Map