Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633662.3 | XP_005633719.1 | 1 - 2 | 634 | 145 | |
| XM_022407836.1 | XP_022263544.1 | 1 - 2 | 634 | 145 | ||
| XM_534051.6 | XP_534051.3 | 1 - 2 | 632 | 145 | ||
More Information
Set Membership: |
|

WEE1
Genomic Map