Assay Details
| Gene Symbol: ZBTB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623049.3 | XP_005623106.1 | 3 | 1460 | 137 | |
| XM_005623052.3 | XP_005623109.1 | 3 | 1351 | 137 | ||
| XM_014115545.2 | XP_013971020.1 | 3 | 1644 | 137 | ||
| XM_014115546.2 | XP_013971021.1 | 3 | 1713 | 137 | ||
| XM_014115547.2 | XP_013971022.1 | 3 | 1566 | 137 | ||
| XM_014115548.2 | XP_013971023.1 | 3 | 1525 | 137 | ||
| XM_022421593.1 | XP_022277301.1 | 3 | 2106 | 137 | ||
| XM_537319.6 | XP_537319.2 | 3 | 1385 | 137 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZBTB14
Genomic Map