Assay Details
| Gene Symbol: PAX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003248.1 | NP_001003248.1 | 6 - 7 | 781 | 134 | |
| GenBank mRNA | X83591.1 | - | 781 | 134 | ||
| X83592.1 | - | 781 | 134 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAX8
Genomic Map