Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417138.1 | XP_022272846.1 | 304 | 149 | ||
| XM_022417144.1 | XP_022272852.1 | 452 | 149 | |||
| XM_022417146.1 | XP_022272854.1 | 577 | 149 | |||
| XM_022417149.1 | XP_022272857.1 | 304 | 149 | |||
| XM_022417151.1 | XP_022272859.1 | 304 | 149 | |||
| XM_854089.4 | XP_859182.1 | 452 | 149 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STX7
Genomic Map