Assay Details
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617120.3 | XP_005617177.1 | 1087 | 125 | ||
| XM_535182.6 | XP_535182.2 | 1059 | 125 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617120.3 | XP_005617177.1 | 1087 | 125 | ||
| XM_535182.6 | XP_535182.2 | 1059 | 125 | |||
Set Membership: |
|