Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617622.2 | XP_005617679.1 | 7 - 8 | 942 | 153 | |
| XM_022411855.1 | XP_022267563.1 | 7 - 8 | 1049 | 153 | ||
| XM_022411857.1 | XP_022267565.1 | 7 - 8 | 909 | 153 | ||
| XM_535324.6 | XP_535324.4 | 6 - 7 | 844 | 153 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

YARS1
Genomic Map