Assay Details
| Gene Symbol: STX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615483.2 | XP_005615540.1 | 290 | 127 | ||
| XM_022417779.1 | XP_022273487.1 | 406 | 127 | |||
| XM_849451.3 | XP_854544.1 | 533 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STX11
Genomic Map