Assay Details
| Gene Symbol: B4GALT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640872.3 | XP_005640929.1 | 6 | 1198 | 152 | |
| XM_545767.6 | XP_545767.3 | 6 | 1175 | 152 | ||
More Information
Set Membership: |
|

B4GALT3
Genomic Map