Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640220.4 | XP_003640268.1 | 3 - 4 | 265 | 112 | |
| XM_005641485.3 | XP_005641542.1 | 3 - 4 | 843 | 112 | ||
| XM_005641486.3 | XP_005641543.1 | 2 - 3 | 309 | 112 | ||
| XM_005641487.3 | XP_005641544.1 | 2 - 3 | 262 | 112 | ||
| XM_005641488.3 | XP_005641545.1 | 2 - 3 | 492 | 112 | ||
| XM_022416052.1 | XP_022271760.1 | 3 - 4 | 432 | 112 | ||
More Information
Set Membership: |
|

CITED1
Genomic Map