Assay Details
| Gene Symbol: SLC1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003136.1 | NP_001003136.1 | 5 - 6 | 688 | 150 | |
| GenBank mRNA | AF067847.1 | - | 5 - 6 | 688 | 150 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC1A3
Genomic Map