Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621659.3 | XP_005621716.1 | 3 - 4 | 672 | 115 | |
| XM_022420208.1 | XP_022275916.1 | 3 - 4 | 676 | 115 | ||
| XM_022420209.1 | XP_022275917.1 | 3 - 4 | 673 | 115 | ||
| XM_022420210.1 | XP_022275918.1 | 2 - 3 | 532 | 115 | ||
| XM_022420211.1 | XP_022275919.1 | 3 - 4 | 678 | 115 | ||
| XM_022420212.1 | XP_022275920.1 | 3 - 4 | 675 | 115 | ||
More Information
Set Membership: |
|

PKMYT1
Genomic Map