Assay Details
| Gene Symbol: MED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253899.1 | NP_001240828.1 | 2 | 381 | 147 | |
| XM_005618858.3 | XP_005618915.1 | 2 | 558 | 147 | ||
| XM_005618859.3 | XP_005618916.1 | 2 | 448 | 147 | ||
| XM_005618861.3 | XP_005618918.1 | 2 | 655 | 147 | ||
| XM_014112761.2 | XP_013968236.1 | 2 | 590 | 147 | ||
| GenBank mRNA | BU749242.1 | - | 2 | 385 | 147 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MED7
Genomic Map