Assay Details
| Gene Symbol: SEPTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435278.4 | XP_003435326.3 | 10 - 11 | 2716 | 103 | |
| XM_005624727.2 | XP_005624784.2 | 7 - 8 | 860 | 103 | ||
| XM_005624729.3 | XP_005624786.1 | 7 - 8 | 806 | 103 | ||
| XM_005624730.2 | XP_005624787.1 | 8 - 9 | 1157 | 103 | ||
| XM_022423199.1 | XP_022278907.1 | 10 - 11 | 2718 | 103 | ||
| XM_022423206.1 | XP_022278914.1 | 10 - 11 | 1028 | 103 | ||
| XM_022423207.1 | XP_022278915.1 | 10 - 11 | 1022 | 103 | ||
| XM_537693.6 | XP_537693.3 | 8 - 9 | 1267 | 103 | ||
| XM_861979.5 | XP_867072.3 | 8 - 9 | 1157 | 103 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEPTIN4
Genomic Map