Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106751.2 | XP_013962226.1 | 2 - 3 | 1071 | 105 | |
| XM_022408779.1 | XP_022264487.1 | 2 - 3 | 1071 | 105 | ||
More Information
Set Membership: |
|

SLC33A1
Genomic Map