Assay Details
| Gene Symbol: PTGDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048107.1 | NP_001041572.1 | 1 | 350 | 85 | |
| XM_005631066.3 | XP_005631123.1 | 3 | 1004 | 85 | ||
| GenBank mRNA | AM231750.1 | - | 1 | 350 | 85 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTGDR2
Genomic Map