Assay Details
| Gene Symbol: KIF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634920.3 | XP_005634977.1 | 2 - 3 | 1624 | 120 | |
| XM_022408855.1 | XP_022264563.1 | 1 - 2 | 1410 | 120 | ||
| XM_022408856.1 | XP_022264564.1 | 2 - 3 | 1581 | 120 | ||
| XM_022408857.1 | XP_022264565.1 | 2 - 3 | 1587 | 120 | ||
| XM_022408858.1 | XP_022264566.1 | 2 - 3 | 1496 | 120 | ||
| XM_022408859.1 | XP_022264567.1 | 2 - 3 | 1593 | 120 | ||
| XM_542954.6 | XP_542954.3 | 2 - 3 | 1593 | 120 | ||
More Information
Set Membership: |
|

KIF3B
Genomic Map