Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636030.3 | XP_005636087.1 | 4 | 1424 | 99 | |
| XM_005636031.3 | XP_005636088.1 | 3 | 1350 | 99 | ||
| XM_005636032.3 | XP_005636089.1 | 3 | 1319 | 99 | ||
| XM_005636034.3 | XP_005636091.1 | 4 | 1423 | 99 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GAL3ST1
Genomic Map