Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424095.1 | XP_022279803.1 | 1820 | 97 | ||
| XM_531646.6 | XP_531646.2 | 1933 | 97 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424095.1 | XP_022279803.1 | 1820 | 97 | ||
| XM_531646.6 | XP_531646.2 | 1933 | 97 | |||
Set Membership: |
|