Assay Details
| Gene Symbol: GTF2H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122658.2 | XP_013978133.1 | 7 - 8 | 1066 | 145 | |
| XM_022407914.1 | XP_022263622.1 | 8 - 9 | 1163 | 145 | ||
More Information
Set Membership: |
|

GTF2H1
Genomic Map