Assay Details
| Gene Symbol: PRKAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630704.3 | XP_005630761.1 | 3 - 4 | 453 | 69 | |
| XM_014120651.2 | XP_013976126.1 | 3 - 4 | 453 | 69 | ||
| XM_014120652.2 | XP_013976127.1 | 3 - 4 | 453 | 69 | ||
| XM_014120653.2 | XP_013976128.1 | 3 - 4 | 453 | 69 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRKAB2
Genomic Map